Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxycarbonylmaleimide 98%

Supplier: AMBEED, INC
Warning

A126245-1G A126245-100G A126245-10G A126245-250MG A126245-1G A126245-5G
103800-778EA 14 USD InStock
103800-778 76718-724 76718-726 76718-728 77312-972 77312-974
N-Methoxycarbonylmaleimide 98%
N-Methoxycarbonylmaleimide
Precaución: for Research Use Only


Formula: C₆H₅NO₄
Boiling Pt: 242.5 °C (760 mm Hg)
Melting Pt: 63…65 °C
Flash Pt: 100.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042755
CAS Number: 55750-48-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty