Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A123328-1G A123328-250MG A123328-100MG
103800-388EA 122 USD InStock
103800-388 77234-222 77745-904
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate 97%
Methyl-2-amino-1H-benzo[d]imidazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉N₃O₂
Boiling Pt: 427 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08752985
CAS Number: 106429-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty