Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-2-cyclopropylacetate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A221797-1G A221797-5G A221797-250MG A221797-1G
103818-944EA 88 USD InStock
103818-944 103849-824 77013-280 77088-424
Methyl-2-amino-2-cyclopropylacetate hydrochloride 97%
Methyl-2-amino-2-cyclopropylacetate hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031219
CAS Number: 535936-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty