Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-amino-6-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A143560-1G A143560-10G A143560-250MG A143560-1G A143560-5G A143560-25G
103803-296EA 36 USD InStock
103803-296 76783-042 76783-044 77299-346 77299-348 77316-574
Methyl 2-amino-6-methylbenzoate 98%
Methyl 2-amino-6-methylbenzoate


Formula: C₉H₁₁NO₂
Boiling Pt: 257.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11505989
CAS Number: 18595-13-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty