Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-bromoisonicotinate 97%

Supplier: AMBEED, INC
Warning

A121229-1G A121229-100G A121229-10G A121229-25G A121229-1G
103800-046EA 6 USD InStock
103800-046 77161-316 77161-318 77161-320 77312-162
Methyl-2-bromoisonicotinate 97%
Methyl 2-bromoisonicotinate
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
Melting Pt: 36…37 °C
Flash Pt: 115.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03791265
CAS Number: 26156-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty