Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloroacetoacetate 95%

Supplier: AMBEED, INC
Danger

A665708-100G A665708-10G A665708-500G A665708-25G A665708-5G
103840-470EA 5 USD InStock
103840-470 76806-486 76806-488 76806-490 76844-668 76844-670
Methyl-2-chloroacetoacetate 95%
Methyl-2-chloroacetoacetate
Precaución: for Research Use Only


Formula: C₅H₇ClO₃
Boiling Pt: 229 °C (760 mm Hg)
Melting Pt: …33……32.7 °C
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008757
CAS Number: 4755-81-1
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty