Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-cyanobenzoate 96%

Supplier: AMBEED, INC
Warning

A184824-5G A184824-10G A184824-250MG A184824-1G A184824-25G A184824-5G
103815-790EA 8 USD InStock
103815-790 103846-356 76720-996 76720-998 77083-980 77301-922 77301-924
Methyl-2-cyanobenzoate 96%
Methyl-2-cyanobenzoate
Precaución: for Research Use Only


Formula: C₉H₇NO₂
Melting Pt: 49…51 °C
Flash Pt: 136.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00016376
CAS Number: 6587-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty