Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-fluoro-4-iodobenzoate 97%

Supplier: AMBEED, INC
Warning

A131517-1G A131517-10G A131517-250MG A131517-5G A131517-1G
103801-594EA 89 USD InStock
103801-594 77149-632 77149-634 77298-494 77314-094
Methyl-2-fluoro-4-iodobenzoate 97%
Methyl 2-fluoro-4-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₆FIO₂
MW: 280.0400 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD11846098
CAS Number: 204257-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty