Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-formylisonicotinate 98%

Supplier: AMBEED, INC
Warning

A215232-1G A215232-5G A215232-25G A215232-1G A215232-5G
103818-466EA 43 USD InStock
103818-466 103849-170 76826-018 77087-692 77087-694
Methyl 2-formylisonicotinate 98%
Methyl 2-formylisonicotinate
Precaución: for Research Use Only


Formula: C₈H₇NO₃
MW: 165.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14582033
CAS Number: 125104-34-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty