Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-hydroxy-6-methoxybenzoate 98%

Supplier: AMBEED, INC
Warning

A335406-1G A335406-5G A335406-100MG A335406-10G A335406-250MG A335406-1G
103825-690EA 49 USD InStock
103825-690 103836-660 76839-674 77015-480 77015-482 77015-484 77306-486
Methyl 2-hydroxy-6-methoxybenzoate 98%
Methyl 2-hydroxy-6-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204264
CAS Number: 22833-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty