Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methylisonicotinate 97%

Supplier: AMBEED, INC
Warning

A231037-1G A231037-5G A231037-1G A231037-250MG
103810-748EA 28 USD InStock
103810-748 103850-494 77089-368 77275-742
Methyl 2-methylisonicotinate 97%
Methyl 2-methylisonicotinate
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.163 g/mol
Boiling Pt: 221.9 °C (760 mm Hg)
Melting Pt: 4…5 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD09991707
CAS Number: 16830-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty