Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3,5-dioxohexanoate 95%

Supplier: AMBEED, INC
Warning

A112055-1G A112055-1G A112055-100MG A112055-250MG
103797-942EA 212 USD InStock
103797-942 103823-774 77284-206 77666-636 77871-454
Methyl 3,5-dioxohexanoate 95%
Methyl 3,5-dioxohexanoate
Precaución: for Research Use Only


Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 221.6 °C (760 mm Hg)
Flash Pt: 89.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12547105
CAS Number: 29736-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty