Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-amino-5-methylthiophene-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A145520-1G A145520-5G A145520-10G A145520-25G A145520-1G A145520-5G
103803-550EA 10 USD InStock
103803-550 103841-634 76719-502 76719-504 77316-936 77316-938
Methyl-3-amino-5-methylthiophene-2-carboxylate 97%
Methyl-3-amino-5-methylthiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉NO₂S
MW: 171.214 g/mol
Boiling Pt: 319.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00130095
CAS Number: 76575-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty