Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-aminofuran-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A170705-1G A170705-5G A170705-100MG A170705-250MG A170705-1G A170705-5G
103806-530EA 63 USD InStock
103806-530 103844-804 76784-220 76784-222 77081-916 77081-918
Methyl-3-aminofuran-2-carboxylate 95%
Methyl-3-aminofuran-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₇NO₃
MW: 141.1247 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953568
CAS Number: 956034-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty