Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-4-chloro-5-fluorobenzoate 95%

Supplier: AMBEED, INC
Warning

A168187-1G A168187-5G A168187-1G A168187-5G
103806-260EA 63 USD InStock
103806-260 103844-518 77081-570 77081-572
Methyl 3-bromo-4-chloro-5-fluorobenzoate 95%
Methyl 3-bromo-4-chloro-5-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11846086
CAS Number: 1160574-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty