Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-ethynylbenzoate 97%

Supplier: AMBEED, INC
Warning

A164558-1G A164558-5G A164558-10G A164558-1G A164558-25G A164558-5G
103805-880EA 14 USD InStock
103805-880 103844-136 76872-150 77320-314 77320-316 77320-318
Methyl 3-ethynylbenzoate 97%
Methyl 3-ethynylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₈O₂
Boiling Pt: 251.1 °C (760 mm Hg)
Melting Pt: 53.5…55 °C
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703579
CAS Number: 10602-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty