Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-fluoro-4-(hydroxymethyl)benzoate 95%

Supplier: AMBEED, INC
Warning

A104742-1G A104742-5G A104742-10G A104742-250MG A104742-1G A104742-5G
103796-770EA 37 USD InStock
103796-770 103822-494 76809-516 76809-518 77309-354 77309-356
Methyl 3-fluoro-4-(hydroxymethyl)benzoate 95%
Methyl 3-fluoro-4-(hydroxymethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD09836511
CAS Number: 937636-18-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty