Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-fluoro-5-hydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A309581-1G A309581-1G A309581-5G A309581-100MG A309581-10G A309581-250MG A309581-25G
103814-592EA 32 USD InStock
103814-592 103835-772 76838-538 76838-540 76988-928 76988-930 76988-932 76988-934
Methyl-3-fluoro-5-hydroxybenzoate 98%
Methyl-3-fluoro-5-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₇FO₃
Boiling Pt: 290.7±25.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD14698043
CAS Number: 1072004-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty