Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-hydroxy-1H-pyrrole-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A103465-1G A103465-5G A103465-250MG A103465-1G A103465-5G
103796-556EA 91 USD InStock
103796-556 103822-262 76982-286 77283-100 77283-102
Methyl 3-hydroxy-1H-pyrrole-2-carboxylate 97%
Methyl 3-hydroxy-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11975599
CAS Number: 79068-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty