Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-hydroxy-2-thiophenecarboxylate 97%

Supplier: AMBEED, INC
Warning

A256292-1G A256292-5G A256292-1G A256292-25G A256292-5G A256292-100G A256292-10G
103812-260EA 7 USD InStock
103812-260 103852-124 77091-614 77091-616 77091-618 77113-568 77113-570
Methyl-3-hydroxy-2-thiophenecarboxylate 97%
Methyl-3-hydroxy-2-thiophenecarboxylate
Precaución: for Research Use Only


Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 232.304 °C (760 mm Hg)
Melting Pt: 38…43 °C
Flash Pt: 94.295 °C
Storage Temperature: Ambient
MDL Number: MFCD00055642
CAS Number: 5118-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty