Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate 97%

Supplier: AMBEED, INC
Warning

A277802-1G A277802-5G A277802-250MG A277802-1G A277802-5G
103813-368EA 14 USD InStock
103813-368 103853-306 77002-090 77305-274 77305-276
Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate 97%
Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate
Precaución: for Research Use Only


Formula: C₁₁H₈Cl₂O₄
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD03001261
CAS Number: 374679-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty