Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(cyanoacetyl)benzoate 96%

Supplier: AMBEED, INC
Warning

A458211-5G A458211-100MG A458211-250MG A458211-1G A458211-5G A458211-25G
103827-886EA 32 USD InStock
103827-886 103838-962 76725-660 76725-662 76842-828 76842-830 77879-999
Methyl 4-(cyanoacetyl)benzoate 96%
Methyl 4-(cyanoacetyl)benzoate
Precaución: for Research Use Only


Formula: C₁₁H₉NO₃
Boiling Pt: 397.7 °C (760 mm Hg)
Melting Pt: 171…173 °C
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00015378
CAS Number: 69316-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty