Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate 97%

Supplier: AMBEED, INC
Warning

A207749-1G A207749-100MG A207749-250MG A207749-1G
103817-818EA 78 USD InStock
103817-818 76825-782 76825-784 77086-788
Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate 97%
Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₈O₃
MW: 198.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20275559
CAS Number: 94994-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty