Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(trifluoromethyl)benzoate 98%

Supplier: AMBEED, INC
Warning

A202153-10G A202153-500G A202153-5G A202153-1000G A202153-25G
103809-366EA 196 USD InStock
103809-366 76999-376 76999-378 76999-380 77086-090 77086-092
Methyl-4-(trifluoromethyl)benzoate 98%
Methyl-4-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₇F₃O₂
Boiling Pt: 199.2 °C (760 mm Hg)
Melting Pt: 13…14 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042324
CAS Number: 2967-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty