Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-amino-3-iodobenzoate 98%

Supplier: AMBEED, INC
Warning

A100023-1G A100023-5G A100023-100G A100023-10G A100023-25G A100023-500G A100023-1G A100023-5G
103795-202EA 6 USD InStock
103795-202 103820-924 77093-448 77093-450 77093-452 77093-454 77296-536 77296-538
Methyl-4-amino-3-iodobenzoate 98%
Methyl 4-amino-3-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₈INO₂
MW: 277.0600 g/mol
Melting Pt: 86…91 °C
Flash Pt: 177.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00090423
CAS Number: 19718-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty