Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-1H-indole-7-carboxylate 95%

Supplier: AMBEED, INC
Warning

A122383-1G A122383-100MG A122383-250MG A122383-1G
103800-216EA 102 USD InStock
103800-216 76983-324 76983-326 77312-350
Methyl 4-bromo-1H-indole-7-carboxylate 95%
Methyl 4-bromo-1H-indole-7-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD20487964
CAS Number: 1224724-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty