Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A177604-1G A177604-5G A177604-100MG A177604-250MG A177604-1G A177604-25G A177604-5G
103807-250EA 34 USD InStock
103807-250 103845-580 77096-696 77096-698 77301-512 77301-514 77301-516
Methyl-4-bromo-3-hydroxythiophene-2-carboxylate 95%
Methyl 4-bromo-3-hydroxythiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅BrO₃S
MW: 237.08 g/mol
Boiling Pt: 247.9±35.0 °C (760 mm Hg)
Melting Pt: 79…80 °C
Flash Pt: 103.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00052081
CAS Number: 95201-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty