Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-bromobenzoate 98%

Supplier: AMBEED, INC
Warning

A215466-100G A215466-10G A215466-500G A215466-1000G A215466-25G
103809-506EA 137 USD InStock
103809-506 103849-198 76814-716 76814-718 76814-720 77087-728 77087-730
Methyl-4-bromobenzoate 98%
Methyl-4-bromobenzoate
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 246
Flash Pt: 112.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00013531
CAS Number: 619-42-1
Merck Index: 13,01392
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty