Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-bromopicolinate 97%

Supplier: AMBEED, INC
Warning

A121174-1G A121174-100G A121174-10G A121174-250MG A121174-25G A121174-500G A121174-1G A121174-5G
103800-042EA 6 USD InStock
103800-042 77161-294 77161-296 77161-298 77161-300 77161-302 77312-158 77312-160
Methyl-4-bromopicolinate 97%
Methyl 4-bromopicolinate
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 285.9 °C (760 mm Hg)
Melting Pt: 54…56 °C
Flash Pt: 126.749 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07374897
CAS Number: 29681-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty