Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-chloro-3-fluoropicolinate 98%

Supplier: AMBEED, INC
Warning

A122400-1G A122400-100MG A122400-250MG A122400-1G
103800-224EA 131 USD InStock
103800-224 77009-480 77009-482 77297-866
Methyl 4-chloro-3-fluoropicolinate 98%
Methyl 4-chloro-3-fluoropicolinate
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
MW: 189.572 g/mol
Boiling Pt: 267.7±40.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD16293406
CAS Number: 1034921-05-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty