Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-Chloro-3-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A263347-1G A263347-5G A263347-100G A263347-1G
103812-616EA 5 USD InStock
103812-616 103852-506 76722-890 77092-144
Methyl-4-Chloro-3-methylbenzoate 97%
Methyl-4-Chloro-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
Boiling Pt: 247 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00151838
CAS Number: 91367-05-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty