Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-iodo-2-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A202441-1G A202441-5G A202441-100MG A202441-250MG A202441-1G
103817-390EA 25 USD InStock
103817-390 103848-030 76814-320 76814-322 77302-814
Methyl 4-iodo-2-methylbenzoate 97%
Methyl 4-iodo-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₉IO₂
Boiling Pt: 285.7±28.0 °C (760 mm Hg)
Flash Pt: 126.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18399464
CAS Number: 103440-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty