Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-iodobenzoate 97%

Supplier: AMBEED, INC
Warning

A210083-5G A210083-25G A210083-100G A210083-10G A210083-500G A210083-5G
103848-666EA 5 USD InStock
103848-666 77087-036 77237-364 77237-366 77237-368 77303-114
Methyl-4-iodobenzoate 97%
Methyl-4-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₇IO₂
MW: 262.05
Boiling Pt: 276
Melting Pt: 112
Density: 2.02
Flash Pt: 120.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00016353
CAS Number: 619-44-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty