Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A101989-1G A101989-5G A101989-100MG A101989-10G A101989-250MG A101989-25G A101989-1G A101989-5G
103796-240EA 18 USD InStock
103796-240 103821-936 76809-276 76809-278 76809-280 76809-282 77308-818 77308-820
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester 95%
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₂₁BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD11520530
CAS Number: 882679-40-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty