Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-oxocyclohexanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A124142-1G A124142-10G A124142-25G A124142-500G A124142-5G A124142-1G
103800-488EA 9 USD InStock
103800-488 76870-402 76870-404 76870-406 77297-970 77312-620
Methyl-4-oxocyclohexanecarboxylate 97%
Methyl-4-oxocyclohexanecarboxylate
Precaución: For research use only.


Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Boiling Pt: 230.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00270324
CAS Number: 6297-22-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty