Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-(bromomethyl)-2-fluorobenzoate 95%

Supplier: AMBEED, INC
Danger

A192839-1G A192839-1G A192839-100MG A192839-250MG
103816-486EA 230 USD InStock
103816-486 77084-982 77236-944 77236-946
Methyl 5-(bromomethyl)-2-fluorobenzoate 95%
Methyl 5-(bromomethyl)-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD25459290
CAS Number: 709-45-5
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty