Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-4-hydroxynicotinate 98%

Supplier: AMBEED, INC
Warning

A302998-1G A302998-5G A302998-100MG A302998-250MG
103814-360EA 181 USD InStock
103814-360 103835-524 76988-814 76988-816
Methyl 5-bromo-4-hydroxynicotinate 98%
Methyl 5-bromo-4-hydroxynicotinate
Precaución: for Research Use Only


Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD26383572
CAS Number: 1529795-43-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty