Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-4-nitrothiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A301080-1G A301080-5G A301080-100MG A301080-250MG A301080-1G A301080-5G
103814-294EA 75 USD InStock
103814-294 103835-444 76772-642 76772-644 77305-820 77305-822
Methyl 5-bromo-4-nitrothiophene-2-carboxylate 95%
Methyl 5-bromo-4-nitrothiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₄BrNO₄S
Boiling Pt: 324.1 °C (760 mm Hg)
Flash Pt: 149.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01567069
CAS Number: 38239-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty