Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-chloropyrazine-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A142392-1G A142392-100G A142392-10G A142392-1G A142392-25G A142392-5G
103803-154EA 10 USD InStock
103803-154 76972-668 76972-670 77316-380 77316-382 77316-384
Methyl-5-chloropyrazine-2-carboxylate 98%
Methyl 5-chloropyrazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₂
Boiling Pt: 242.8 °C (760 mm Hg)
Flash Pt: 100.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01632102
CAS Number: 33332-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty