Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-fluoro-2-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A179057-1G A179057-5G A179057-1G A179057-25G A179057-5G A179057-100G A179057-10G
103807-392EA 9 USD InStock
103807-392 103845-732 77083-176 77083-178 77083-180 77111-338 77111-340
Methyl-5-fluoro-2-methylbenzoate 98%
Methyl 5-fluoro-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₉FO₂
MW: 168.1700 g/mol
Boiling Pt: 211.9 °C (760 mm Hg)
Flash Pt: 79.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00221457
CAS Number: 175278-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty