Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-amino-4-chloronicotinate 96%

Supplier: AMBEED, INC
Warning

A272416-1G A272416-100MG A272416-250MG
103813-096EA 684 USD InStock
103813-096 103853-008 77238-776 77238-778
Methyl 6-amino-4-chloronicotinate 96%
Methyl 6-amino-4-chloronicotinate
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
MW: 186.59 g/mol
Boiling Pt: 335.4 °C (760 mm Hg)
Flash Pt: 156.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18250740
CAS Number: 1260666-60-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty