Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-amino-5-bromopicolinate 97%

Supplier: AMBEED, INC
Warning

A766524-1G A766524-5G A766524-100MG A766524-250MG
103840-916EA 328 USD InStock
103840-916 76845-152 76845-154 77036-212 77036-214
Methyl-6-amino-5-bromopicolinate 97%
Methyl 6-amino-5-bromopicolinate
Precaución: for Research Use Only


Formula: C₇H₇BrN₂O₂
MW: 231.0500 g/mol
Boiling Pt: 359.186 °C (760 mm Hg)
Melting Pt: 142…146 °C
Flash Pt: 171.03 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062950
CAS Number: 178876-82-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty