Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-aminohexanoate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A109019-1G A109019-100G A109019-10G A109019-250MG A109019-25G A109019-1G A109019-5G A109019-500G
103797-428EA 6 USD InStock
103797-428 103823-216 76717-972 76717-974 76717-976 76717-978 77283-796 77310-046 77666-586
Methyl-6-aminohexanoate hydrochloride 98%
Methyl-6-aminohexanoate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₆ClNO₂
Melting Pt: 117…124 °C
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035455
CAS Number: 1926-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty