Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromo-1H-indazole-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A164500-1G A164500-5G A164500-10G A164500-250MG A164500-1G A164500-25G A164500-5G
103805-870EA 21 USD InStock
103805-870 103844-130 77011-362 77011-364 77320-306 77320-308 77320-310
Methyl-6-bromo-1H-indazole-3-carboxylate 95%
Methyl 6-bromo-1H-indazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₇BrN₂O₂
Boiling Pt: 399.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07371573
CAS Number: 885278-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty