Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-bromo-4-chloronicotinate 96%

Supplier: AMBEED, INC
Warning

A105225-1G A105225-5G A105225-250MG A105225-5G A105225-1G
103796-858EA 94 USD InStock
103796-858 103822-586 77160-460 77283-348 77309-460
Methyl 6-bromo-4-chloronicotinate 96%
Methyl 6-bromo-4-chloronicotinate
Precaución: for Research Use Only


Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Melting Pt: 63…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18257847
CAS Number: 1256789-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty