Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromobenzo[b]thiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A147824-1G A147824-5G A147824-10G A147824-250MG A147824-1G A147824-25G A147824-5G
103803-838EA 18 USD InStock
103803-838 103841-946 77010-622 77010-624 77317-354 77317-356 77317-358
Methyl-6-bromobenzo[b]thiophene-2-carboxylate 95%
Methyl 6-bromobenzo[b]thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₇BrO₂S
Boiling Pt: 358.9 °C (760 mm Hg)
Flash Pt: 170.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371541
CAS Number: 360576-01-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty