Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromopicolinate 98%

Supplier: AMBEED, INC
Warning

A131301-1G A131301-100G A131301-10G A131301-500G A131301-1G A131301-25G A131301-5G
103801-562EA 9 USD InStock
103801-562 77094-980 77094-982 77094-984 77314-056 77314-058 77314-060
Methyl-6-bromopicolinate 98%
Methyl 6-bromopicolinate
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 289.7 °C (760 mm Hg)
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD06203934
CAS Number: 26218-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty