Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-chloro-2-methylnicotinate 97%

Supplier: AMBEED, INC
Warning

A781055-5G A781055-100MG A781055-250MG A781055-1G A781055-5G
103840-980EA 245 USD InStock
103840-980 77244-088 77244-090 77308-322 77308-324
Methyl 6-chloro-2-methylnicotinate 97%
Methyl 6-chloro-2-methylnicotinate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Boiling Pt: 235.842 °C (760 mm Hg)
Flash Pt: 96.434 °C
Storage Temperature: Ambient
MDL Number: MFCD14155903
CAS Number: 851759-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty