Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-chloro-3-methylpicolinate 98%

Supplier: AMBEED, INC
Warning

A118132-1G A118132-5G A118132-10G A118132-250MG A118132-5G
103798-996EA 47 USD InStock
103798-996 103824-888 77026-242 77026-244 77311-678
Methyl-6-chloro-3-methylpicolinate 98%
Methyl 6-chloro-3-methylpicolinate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
Flash Pt: 128.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12756112
CAS Number: 878207-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty