Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid methyl ester 97%

Supplier: AMBEED, INC
Warning

A262980-1G A262980-5G A262980-10G A262980-250MG A262980-1G A262980-5G
103812-592EA 17 USD InStock
103812-592 103852-488 76771-926 76771-928 77092-118 77304-862
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid methyl ester 97%
6-Chloroimidazo[1,2-b]pyridazine-2-carboxylic acid methyl ester
Precaución: for Research Use Only


Formula: C₈H₆ClN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11044753
CAS Number: 572910-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty